Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414991.1 | XP_022270699.1 | 6 - 7 | 1389 | 98 | |
| XM_545569.6 | XP_545569.3 | 6 - 7 | 1392 | 98 | ||
More Information
Set Membership: |
|

NAB1
Genomic Map