Assay Details
| Gene Symbol: BLZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622551.3 | XP_005622608.1 | 2 - 3 | 584 | 73 | |
| XM_547475.6 | XP_547475.2 | 3 - 4 | 655 | 73 | ||
More Information
Set Membership: |
|

BLZF1
Genomic Map