Assay Details
| Gene Symbol: PDE6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003156.1 | NP_001003156.1 | 3 | 290 | 94 | |
| XM_022409540.1 | XP_022265248.1 | 3 | 248 | 94 | ||
| GenBank mRNA | AF109151.1 | - | 3 | 290 | 94 | |
| AY029186.1 | - | 3 | 253 | 94 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PDE6D
Genomic Map