Assay Details
| Gene Symbol: B3GAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631619.3 | XP_005631676.1 | 3 - 4 | 809 | 63 | |
| XM_540906.6 | XP_540906.2 | 3 - 4 | 805 | 63 | ||
More Information
Set Membership: |
|

B3GAT3
Genomic Map