Assay Details
| Gene Symbol: REXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633123.3 | XP_005633180.1 | 14 - 15 | 4025 | 152 | |
| XM_014122211.2 | XP_013977686.1 | 15 - 16 | 3448 | 152 | ||
| XM_022407268.1 | XP_022262976.1 | 16 - 17 | 4154 | 152 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

REXO1
Genomic Map