Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407035.1 | XP_022262743.1 | 4 | 390 | 113 | |
| XM_022407036.1 | XP_022262744.1 | 3 | 362 | 113 | ||
| XM_022407037.1 | XP_022262745.1 | 3 | 275 | 113 | ||
| XM_533912.4 | XP_533912.2 | 3 | 469 | 113 | ||
More Information
Set Membership: |
|

TMEM205
Genomic Map