Assay Details
| Gene Symbol: DEDD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415435.1 | XP_022271143.1 | 4 - 5 | 618 | 119 | |
| XM_022415436.1 | XP_022271144.1 | 4 - 5 | 557 | 119 | ||
| XM_022415437.1 | XP_022271145.1 | 4 - 5 | 1142 | 119 | ||
| XM_022415438.1 | XP_022271146.1 | 3 - 4 | 679 | 119 | ||
| XM_022415439.1 | XP_022271147.1 | 3 - 4 | 537 | 119 | ||
More Information
Set Membership: |
|

DEDD
Genomic Map