Assay Details
| Gene Symbol: APOA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619731.3 | XP_005619788.1 | 812 | 111 | ||
| XM_022418257.1 | XP_022273965.1 | 633 | 111 | |||
| XM_546511.6 | XP_546511.1 | 805 | 111 | |||
More Information
Set Membership: |
|

APOA5
Genomic Map