Assay Details
| Gene Symbol: Lrrc51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282002.1 | NP_001268931.1 | 4 - 5 | 504 | 76 | |
| NM_001282003.1 | NP_001268932.1 | 3 - 4 | 441 | 76 | ||
| GenBank mRNA | JI877748.1 | - | 2 - 3 | 349 | 76 | |
| JP062696.1 | - | 2 - 3 | 349 | 76 | ||

Lrrc51
Genomic Map