Assay Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246736.1 | NP_001233665.1 | 1 - 2 | 81 | 61 | |
| GenBank mRNA | JI878699.1 | - | 1 - 2 | 96 | 61 | |
| JP046069.1 | - | 1 - 2 | 96 | 61 | ||
| U05959.1 | - | 1 - 2 | 81 | 61 | ||
More Information
Set Membership: |
|

Pex19
Genomic Map