Assay Details
| Gene Symbol: Sepp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256878.1 | NP_001243807.1 | 2 - 3 | 379 | 60 | |
| GenBank mRNA | JI884822.1 | - | 2 - 3 | 379 | 60 | |
| JP052462.1 | - | 2 - 3 | 379 | 60 | ||
More Information
Set Membership: |
|

Sepp1
Genomic Map