Assay Details
| Gene Symbol: Nceh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640560.1 | XP_007638750.1 | 1 - 2 | 340 | 63 | |
| GenBank mRNA | JP062118.1 | - | 1 - 2 | 210 | 63 | |
More Information
Set Membership: |
|

Nceh1
Genomic Map