Assay Details
| Gene Symbol: Pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512349.2 | XP_003512397.1 | 5 - 6 | 812 | 68 | |
| XM_007652840.1 | XP_007651030.1 | 4 - 5 | 693 | 68 | ||
| XM_007652841.1 | XP_007651031.1 | 3 - 4 | 369 | 68 | ||
More Information
Set Membership: |
|

Pcmtd2
Genomic Map