Assay Details
| Gene Symbol: LOC100754077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511069.2 | XP_003511117.2 | 4 - 5 | 521 | 83 | |
| GenBank mRNA | JI876717.1 | - | 2754 | 83 | ||
| JP038497.1 | - | 2755 | 83 | |||
More Information
Set Membership: |
|

LOC100754077
Genomic Map