Assay Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505530.2 | XP_003505578.1 | 3 - 4 | 503 | 64 | |
| XM_007646022.1 | XP_007644212.1 | 3 - 4 | 503 | 64 | ||
| XM_007646023.1 | XP_007644213.1 | 2 - 3 | 299 | 64 | ||
More Information
Set Membership: |
|

Fmo2
Genomic Map