Assay Details
| Gene Symbol: LOC100758234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513939.1 | XP_003513987.1 | 1 - 2 | 168 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100758234| Gene Symbol: LOC100758234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513939.1 | XP_003513987.1 | 1 - 2 | 168 | 73 | |
Set Membership: |
|