Assay Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512994.2 | XP_003513042.1 | 1 - 2 | 525 | 62 | |
| GenBank mRNA | JI875441.1 | - | 1 - 2 | 492 | 62 | |
| JP043957.1 | - | 1 - 2 | 492 | 62 | ||
More Information
Set Membership: |
|

Kdelc1
Genomic Map