Assay Details
| Gene Symbol: LOC100757058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510509.2 | XP_003510557.2 | 3 - 4 | 1482 | 89 | |
| GenBank mRNA | JI874974.1 | - | 3 - 4 | 333 | 89 | |
| JP057599.1 | - | 2 - 3 | 270 | 89 | ||
More Information
Set Membership: |
|

LOC100757058
Genomic Map