Assay Details
| Gene Symbol: Batf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502770.2 | XP_003502818.1 | 2 - 3 | 397 | 70 | |
| GenBank mRNA | JI862723.1 | - | 2 - 3 | 393 | 70 | |
| JP059757.1 | - | 2 - 3 | 391 | 70 | ||
More Information
Set Membership: |
|

Batf
Genomic Map