Assay Details
| Gene Symbol: Snai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502472.2 | XP_003502520.1 | 2 - 3 | 738 | 91 | |
| GenBank mRNA | JI869057.1 | - | 1 - 2 | 132 | 91 | |
More Information
Set Membership: |
|

Snai1
Genomic Map