Assay Details
| Gene Symbol: Tsc22d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641023.1 | XP_007639213.1 | 2 | 3106 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsc22d1| Gene Symbol: Tsc22d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641023.1 | XP_007639213.1 | 2 | 3106 | 67 | |
Set Membership: |
|