Assay Details
| Gene Symbol: RCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059291.1 | XP_017914780.1 | 3 - 4 | 1045 | 72 | |
| GenBank mRNA | JO497545.2 | - | 3 - 4 | 718 | 72 | |
More Information
Set Membership: |
|

RCN1
Genomic Map