Assay Details
| Gene Symbol: SLC2A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060898.1 | XP_017916387.1 | 4 - 5 | 996 | 80 | |
| XM_018060899.1 | XP_017916388.1 | 4 - 5 | 996 | 80 | ||
| XM_018060900.1 | XP_017916389.1 | 4 - 5 | 583 | 80 | ||
| XM_018060901.1 | XP_017916390.1 | 2 - 3 | 1371 | 80 | ||
| XM_018060902.1 | XP_017916391.1 | 4 - 5 | 996 | 80 | ||
| XM_018060903.1 | XP_017916392.1 | 4 - 5 | 996 | 80 | ||
| XM_018060904.1 | XP_017916393.1 | 4 - 5 | 996 | 80 | ||
| XM_018060905.1 | XP_017916394.1 | 4 - 5 | 996 | 80 | ||
| XM_018060906.1 | XP_017916395.1 | 4 - 5 | 996 | 80 | ||
| GenBank mRNA | JO427609.1 | - | 8 - 9 | 1844 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018060901.1 transcripts.
Set Membership: |
|

SLC2A11
Genomic Map