Assay Details
| Gene Symbol: ZNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063543.1 | XP_017919032.1 | 3 - 4 | 648 | 94 | |
| XM_018063544.1 | XP_017919033.1 | 3 - 4 | 412 | 94 | ||
| GenBank mRNA | JT675507.1 | - | 3 - 4 | 378 | 94 | |
More Information
Set Membership: |
|

ZNF8
Genomic Map