Assay Details
| Gene Symbol: SLC25A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042040.1 | XP_017897529.1 | 3 - 4 | 1078 | 61 | |
| GenBank mRNA | JI861760.1 | - | 3 - 4 | 738 | 61 | |
| JO634330.2 | - | 1 - 2 | 551 | 61 | ||
More Information
Set Membership: |
|

SLC25A4
Genomic Map