Assay Details
| Gene Symbol: LCLAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055477.1 | XP_017910966.1 | 4 - 5 | 661 | 80 | |
| XM_018055478.1 | XP_017910967.1 | 3 - 4 | 492 | 80 | ||
| GenBank mRNA | JT762624.1 | - | 3 - 4 | 410 | 80 | |
More Information
Set Membership: |
|

LCLAT1
Genomic Map