Assay Details
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695578.3 | XP_005695635.2 | 2 - 3 | 580 | 56 | |
XM_013973347.2 | XP_013828801.2 | 2 - 3 | 664 | 56 | ||
XM_018066822.1 | XP_017922311.1 | 2 - 3 | 391 | 56 | ||
GenBank mRNA | JO601607.2 | - | 6 - 7 | 1299 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
