Assay Details
| Gene Symbol: CADM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046350.1 | XP_017901839.1 | 6 - 7 | 961 | 61 | |
| XM_018046351.1 | XP_017901840.1 | 5 - 6 | 851 | 61 | ||
| GenBank mRNA | JO633959.2 | - | 4 - 5 | 830 | 61 | |
| KA309326.1 | - | 4 - 5 | 604 | 61 | ||
More Information
Set Membership: |
|

CADM3
Genomic Map