Assay Details
| Gene Symbol: PPP1R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679168.3 | XP_005679225.2 | 1 - 2 | 814 | 119 | |
| XM_013963316.2 | XP_013818770.2 | 2 - 3 | 503 | 119 | ||
More Information
Set Membership: |
|

PPP1R3A
Genomic Map