Assay Details
| Gene Symbol: AGPAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684957.3 | XP_005685014.2 | 7 - 8 | 1092 | 64 | |
| XM_005684958.3 | XP_005685015.2 | 7 - 8 | 1179 | 64 | ||
| XM_005684959.3 | XP_005685016.2 | 7 - 8 | 1056 | 64 | ||
| GenBank mRNA | JO621528.2 | - | 7 - 8 | 1151 | 64 | |
More Information
Set Membership: |
|

AGPAT4
Genomic Map