Assay Details
| Gene Symbol: EIF2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691356.3 | XP_005691413.1 | 4 - 5 | 486 | 63 | |
| GenBank mRNA | JR138766.1 | - | 1168 | 63 | ||
More Information
Set Membership: |
|

EIF2B1
Genomic Map