Assay Details
| Gene Symbol: KRT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693818.3 | XP_005693875.3 | 1 - 2 | 721 | 82 | |
| GenBank mRNA | KA321729.1 | - | 1 - 2 | 623 | 82 | |
More Information
Set Membership: |
|

KRT14
Genomic Map