Assay Details
| Gene Symbol: EDIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683400.3 | XP_005683457.2 | 8 - 9 | 1058 | 74 | |
| XM_005683401.3 | XP_005683458.2 | 7 - 8 | 993 | 74 | ||
| GenBank mRNA | JO644585.2 | - | 3 - 4 | 695 | 74 | |
More Information
Set Membership: |
|

EDIL3
Genomic Map