Assay Details
| Gene Symbol: C19H17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064536.1 | XP_017920025.1 | 3 - 4 | 456 | 107 | |
| XM_018064537.1 | XP_017920026.1 | 3 - 4 | 456 | 107 | ||
| GenBank mRNA | JO351749.2 | - | 3 - 4 | 552 | 107 | |
| JO611484.2 | - | 3 - 4 | 334 | 107 | ||
| KA318145.1 | - | 3 - 4 | 399 | 107 | ||
More Information
Set Membership: |
|

C19H17orf49
Genomic Map