Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682380.3 | XP_005682437.1 | 3 - 4 | 350 | 85 | |
| XM_005682381.3 | XP_005682438.1 | 3 - 4 | 381 | 85 | ||
| XM_005682382.3 | XP_005682439.1 | 3 - 4 | 411 | 85 | ||
| GenBank mRNA | JO420562.2 | - | 3 - 4 | 361 | 85 | |
More Information
Set Membership: |
|

TMEM205
Genomic Map