Assay Details
| Gene Symbol: ZNF354A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050558.1 | XP_017906047.1 | 4 - 5 | 1339 | 115 | |
| XM_018050560.1 | XP_017906049.1 | 4 - 5 | 816 | 115 | ||
| XM_018050561.1 | XP_017906050.1 | 4 - 5 | 1261 | 115 | ||
| XM_018050562.1 | XP_017906051.1 | 4 - 5 | 502 | 115 | ||
| GenBank mRNA | JT659514.1 | - | 1 - 2 | 715 | 115 | |
More Information
Set Membership: |
|

ZNF354A
Genomic Map