Assay Details
| Gene Symbol: LOC102190981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690612.3 | XP_005690669.2 | 2 - 3 | 231 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102190981| Gene Symbol: LOC102190981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690612.3 | XP_005690669.2 | 2 - 3 | 231 | 91 | |
Set Membership: |
|