Assay Details
| Gene Symbol: GDPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009390.2 | XP_009007638.2 | 5 - 6 | 640 | 80 | |
| XM_017978842.1 | XP_017834331.1 | 5 - 6 | 640 | 80 | ||
More Information
Set Membership: |
|

GDPD3
Genomic Map