Assay Details
| Gene Symbol: LSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746361.4 | XP_002746407.1 | 2 - 3 | 292 | 74 | |
| XM_017970975.1 | XP_017826464.1 | 2 - 3 | 257 | 74 | ||
More Information
Set Membership: |
|

LSM2
Genomic Map