Assay Details
| Gene Symbol: GPRC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755946.3 | XP_002755992.1 | 2 - 3 | 1254 | 59 | |
| XM_009009257.2 | XP_009007505.1 | 2 - 3 | 1426 | 59 | ||
More Information
Set Membership: |
|

GPRC5B
Genomic Map