Assay Details
| Gene Symbol: FGF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745674.4 | XP_002745720.1 | 2 - 3 | 733 | 109 | |
| XM_008993100.2 | XP_008991348.1 | 2 - 3 | 408 | 109 | ||
More Information
Set Membership: |
|

FGF5
Genomic Map