Assay Details
| Gene Symbol: ZNF131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745012.3 | XP_002745058.1 | 5 - 6 | 847 | 98 | |
| XM_008992108.2 | XP_008990356.1 | 5 - 6 | 1193 | 98 | ||
More Information
Set Membership: |
|

ZNF131
Genomic Map