Assay Details
| Gene Symbol: ACVR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752485.2 | XP_002752531.1 | 8 - 9 | 1499 | 66 | |
| XM_017976067.1 | XP_017831556.1 | 9 - 10 | 1616 | 66 | ||
More Information
Set Membership: |
|

ACVR1B
Genomic Map