Assay Details
| Gene Symbol: LOC100402183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743935.3 | XP_002743981.3 | 11 - 12 | 3494 | 94 | |
| XM_008981708.1 | XP_008979956.1 | 11 - 12 | 3067 | 94 | ||
More Information
Set Membership: |
|

LOC100402183
Genomic Map