Assay Details
| Gene Symbol: TSC22D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983672.2 | XP_008981920.1 | 4 - 5 | 2549 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSC22D2| Gene Symbol: TSC22D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983672.2 | XP_008981920.1 | 4 - 5 | 2549 | 104 | |
Set Membership: |
|