Assay Details
| Gene Symbol: TMPPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965928.1 | XP_017821417.1 | 14 - 15 | 2476 | 89 | |
| GenBank mRNA | EF214840.1 | - | 1 - 2 | 121 | 89 | |
More Information
Set Membership: |
|

TMPPE
Genomic Map