Assay Details
| Gene Symbol: RNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003971.2 | XP_009002219.1 | 3 - 4 | 484 | 75 | |
| XM_009003972.2 | XP_009002220.1 | 3 - 4 | 408 | 75 | ||
| XM_009003973.2 | XP_009002221.1 | 3 - 4 | 621 | 75 | ||
| XM_009003975.2 | XP_009002223.1 | 3 - 4 | 321 | 75 | ||
| XM_017976139.1 | XP_017831628.1 | 4 - 5 | 539 | 75 | ||
| XM_017976140.1 | XP_017831629.1 | 2 - 3 | 306 | 75 | ||
| XM_017976141.1 | XP_017831630.1 | 4 - 5 | 590 | 75 | ||
More Information
Set Membership: |
|

RNF41
Genomic Map