Assay Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988198.2 | XP_008986446.1 | 3 - 4 | 518 | 85 | |
| XM_008988199.2 | XP_008986447.1 | 3 - 4 | 522 | 85 | ||
| XM_008988201.2 | XP_008986449.1 | 3 - 4 | 660 | 85 | ||
More Information
Set Membership: |
|

ZNF180
Genomic Map