Assay Details
| Gene Symbol: ASIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749817.3 | XP_002749863.1 | 1 - 2 | 1336 | 73 | |
More Information
Set Membership: |
|
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASIC4| Gene Symbol: ASIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749817.3 | XP_002749863.1 | 1 - 2 | 1336 | 73 | |
Set Membership: |
|