Assay Details
| Gene Symbol: PPP1R14D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753532.3 | XP_002753578.1 | 4 | 963 | 100 | |
| XM_009005540.2 | XP_009003788.1 | 5 | 1138 | 100 | ||
More Information
Set Membership: |
|

PPP1R14D
Genomic Map